BS 5609 Marine Label Tapes for BradyJet™ J7300 Color Label Printer
Supplier: Brady Worldwide
170469 170470 176619 176620 176586
77581-050RL 411.28 CAD
77581-050 77580-922 77581-104 77581-106 77581-076
BS 5609 Marine Label Tapes for BradyJet™ J7300 Color Label Printer
Labels
Labels are BS 5609 Sections 2 and 3 certified to ensure identification stays compliant, stuck, and seen during marine transport. 
  • Permanent acrylic adhesive
  • Matte finish
  • Service temperature range: –40 to +140 °F
Ideal for GHS, hazardous materials, drum labeling, marine, transportation (DOT), storage, and pharmaceutical labeling. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 colour Label Printer. InkMore Product Information