PermaShield™ Matte Polyester Labels with Overlaminate for BradyJet™ J7300 Color Label Printer
Supplier: Brady Worldwide
176652 176653 176649 176650 176651
77581-022RL 916.81 CAD
77581-022 77581-098 77581-058 77580-960 77580-980
PermaShield™ Matte Polyester Labels with Overlaminate for BradyJet™ J7300 Color Label Printer
Labels
Upgrade the durability and longevity of identification projects, even in the toughest environments, with PermaShield™ polyester labels designed for BradyJet™ printers.
  • Permanent acrylic adhesive
  • Matte finish
  • Chemical-resistant material
  • Service temperature range: –40 to +140 °F
PermaShield™ by Brady is a 2-in-1 package that includes a printable label plus matching clear polyester overlaminate. Simply print label, adhere to surface, and apply the matching overlaminate. Protects against numerous harsh solvents and provides a more glossy appearance.More Product Information