BradyJet™ J7300 Color Label Printer WiFi with Safety and Facility ID Software
Supplier: Brady Worldwide
176877 176878
77581-068EA 3779.68 CAD
77581-068 77580-896
BradyJet™ J7300 Color Label Printer WiFi with Safety and Facility ID Software
Printers Label Printers
Unleash true color with the BradyJet™ J7300 Color label printer. Create vibrant, industrial-grade inkjet for safety labels, signs, and tags in full color. Non-WiFi option available.
  • Full color printing capability
  • PC connected
  • Print resolution: 4800 dpi
  • 2 year warranty
  • Maximum label width: 4.25"
  • Maximum labels per day: 5000
Ideal for industrial safety labeling those changes frequently, from Lean/5S, pipe marking and lockout tagout to OSHA/ANSI signage and custom labels. LabelSense™ technology enables printer, materials, and inks to talk to each other for automatic part recognition, setup, and adjustments. Dual CMY and true black (K) ink cartridges deliver vibrant colors and deeper darks for safety labels that demand attention. Prints smear-free inks on labels that resist water, chemicals, and abrasions for up to two-year outdoor durability (even longer indoors). Print Utility dashboard helps users plan, budget, and troubleshoot with status alerts, a job cost calculator, remote monitoring, and more. Exterior LED lighting and clear cover communicate real-time printer status.
Brady Workstation Safety and Facility ID Suite helps create and print using pre-made templates, graphics, and best-match regulatory color palettes. Optimized Windows® driver allows connection to third-party software and print PDFs.