Polyester Label Tapes for BradyJet™ J7300 Color Label Printer
Supplier: Brady Worldwide
176577 176578 170452 170453 170454 176600 176624 176625
77580-928RL 313.57 CAD
77580-928 77580-926 77580-902 77581-064 77580-898 77580-954 77580-890 77581-048
Polyester Label Tapes for BradyJet™ J7300 Color Label Printer
Labels
General-purpose polyester labels from Brady feature a specially formulated topcoat for improved durability, color, and appearance.
  • Permanent acrylic adhesive
  • Matte finish
  • Durable, chemical-resistant material
  • Suitable for harsh environment, outdoor use
  • Service temperature range: −40 to 140 °F
Inkjet-receptive polyester material showcases vibrant colors, sharp text, and precise graphics. Adheres strongly to smooth surfaces for a wide range of applications, and is durable up to two years outdoors (even longer indoors). Ideal for Lean/5S, general facilityMore Product Information