UL Approved Polyester Label Tapes for BradyJet™ J7300 Color Label Printer
Supplier: Brady Worldwide
176576 176596 176597 176598 176599 176591 176592 176593 176594 176595 176574 176575
77581-086RL 597.61 CAD
77581-086 77581-000 77580-982 77581-062 77580-986 77580-946 77580-916 77581-054 77581-096 77580-950 77580-956 77581-072
UL Approved Polyester Label Tapes for BradyJet™ J7300 Color Label Printer
Labels
Polyester material meets UL 969 Standard for Marking and Labeling and is ideal for product labeling, logos, Right to Know labels, and barcode applications.
  • Permanent acrylic adhesive
  • Matte finish
  • Chemical-resistant material
  • Service temperature range: −40 to 140 °F
Inkjet receptive labels allow users to create labels with variable data and color components in-house. Durable and resistant to abrasions, chemicals, oils, and solvents. Strong acrylic adhesive works best on flat, smooth surfaces. Built-in LabelSense™ technology enables cartridge and printerMore Product Information