AccuTec Pro® Single Edge Blades, Unwrapped
Supplier: Accutec Blades
APBL-7008-0000 APBL-7059-0000
77587-130CS 1607.07 CAD
77587-130 77587-120
AccuTec Pro® Single Edge Blades, Unwrapped
Razors and Blades
AccuTec Pro® Single edge blades feature precision ground edges for sharpness and durability. Blades are unwrapped for high volume use.
  • Bulk, unwrapped blades are ideal for high volume use
Choose 3-facet blades for increased sharpness, or 2-facet blades for increased durability.
Packaging: Each box contains 240 blades.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.