Kinase Dilution Buffers X
Supplier: Sino Biological
K20-09-01
K20-09-05
K20-09-20
77558-906EA
26.86
CAD
77558-906
77558-908
77558-910
Kinase Dilution Buffers X
Buffers
Assay Buffers
The working solution used for diluting kinase proteins.
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.