Phosphatase Assay Buffers II
Supplier: Sino Biological
P02-09-01 P02-09-05
77560-892EA 44.24 CAD
77560-892 77560-894
Phosphatase Assay Buffers II
Buffers Assay Buffers
Stock solution of buffer used for assaying phosphatase protein.
  • Frozen solution of buffer containing: 250 mM Imidazole, pH 7.2
  • Buffer passed through a 0.2 µ filter membrane and packaged aseptically
  • Store product at −20 °C
For optimal storage, aliquot buffer into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.