Showa XC510 Cut-Resistant Industrial Gloves
Supplier: Showa
XC510L-08 XC510M-07 XC510S-06 XC510XL-09 XC510XXL-10
77575-158CS 869.38 CAD
77575-158 77575-150 77575-102 77575-124 77575-092
Showa XC510 Cut-Resistant Industrial Gloves
Gloves
The ultra-light XC510 industrial glove combines an 18-gauge UHMWPE cut-resistant liner with a durable, abrasive-resistant polyurethane palm coating for use in a wide variety of industrial applications.
  • Durable, form-fitting, flexible, and lightweight
  • Cut-resistance level D/A4
  • HPPE reinforced
  • Polyurethane palm coating
Flexible and breathable XC510 industrial gloves provide increased dexterity in a form-fitting, seamless knit design. These gloves are recommended for use in maintenance, glass/window repair, component assembly, engineering, and mechanical industries.
Certifications: Meets ANSI level A4. EN 388:2016+A1:2018, EN ISO 21420:2020. Cat. II.
Packaging: Gloves are packaged 12 pair per polybag, 6 polybags per case (72 total pairs).