Showa AP800 General-Purpose Industrial Gloves
Supplier: Showa
AP800M-07 AP800S-06 AP800XL-09 AP800XXL-10 AP800L-08
77575-098CS 471.3 CAD
77575-098 77575-110 77575-100 77575-172 77575-118
Showa AP800 General-Purpose Industrial Gloves
Gloves
The Showa AP800 industrial glove has a 13-gauge seamless knit with a Microfiber polyester static dissipative yarn and an embossed microporous foam nitrile palm dip.
  • Embossed microporous nitrile palm coating
  • Lightweight, form-fitting, and abrasion-resistant
  • Seamless knit
  • Touchscreen compatible
  • Anti-static liner
  • Latex-free, silicone-free, and fingerprint-free
Certified under the EN 16350 standard, the AP800 industrial glove meets the requirements for electrostatic properties (ESD). This certification ensures the glove’s safe usage in ATEX zones (working area with risk of explosion and/or high flammability). This durable yet breathable glove is recommended for use in automotive, electronics, and mechanical industries.
Certifications: Meets ANSI level A4. EN 388:2016+A1:2018, EN 16350:2014. Cat. II.
Packaging: Gloves are packaged 12 per polybag, 12 polybags per case (144 total pairs).