Moisture Analyzers, MB32, MB62 and MB92
Supplier: Ohaus
30971928 30971929 30971930
77578-404EA 5500.79 CAD
77578-404 77578-406 77578-408
Moisture Analyzers, MB32, MB62 and MB92
Analysers Moisture Analysers
The next generation of MB series moisture analyzers come in a sleek design offering improved durability and stability in performance, with an upgraded loadcell and carbon fiber heating technology.
  • MB32: Ideal for routine moisture testing, providing consistent performance for everyday laboratory and production needs
  • MB62: Versatile and suitable for a variety of mositure applicions across different industries
  • MB92: More advanced option for users requiring a high level of performance and precision in moisture analysis
Newly designed housing with improved ergonomics, updated aesthethics, easy cleaning and maintenance, enhanced durability.
Innovative carbon fiber heating with faster results, energy saving and uniform heating.
Upgraded loadcell contributes to more reliable, precise, and efficient moisture analysis which has durability and reliability, superior overload
protection and enhanced stability.
Increased flexibility with customizable parameters, multiple methods storage, advanced programming and user friendly interface.