Capillary Electrophoresis, BioPhase 8800 System
Supplier: AB SCIEX LLC CA
5089278 5083590
CA76645-504EA 260254.22 CAD
CA76645-504 CA76645-502
Capillary Electrophoresis, BioPhase 8800 System
Electrophoresis Systems Capillary Electrophoresis Systems
The BioPhase 8800 system is a multi-capillary electrophoresis system that enables biopharma scientists to run multiple samples in parallel, accelerating the development and execution of sensitive, high-throughput analytical methods. The increased throughput and reproducibility of the system minimizes bottlenecks in the drug development pipeline, helping shorten time to market.
  • Take on multiple molecules and multiple applications in less time - all on a single platform
  • Shorten time to market with high-throughput, eight-capillary analytics
  • With user-friendly
More Product Information