HPLC Basic Waste Bundles, V2.0

Supplier: SCAT AMERICAS, INC.
75200 750201
77553-658EA 503.25 CAD
77553-658 77553-660
HPLC Basic Waste Bundles, V2.0
Chromatography Supplies Chromatography Spare Parts
HPLC Basic waste bundle.

  • S 60/61 thread
  • HDPE material

HPLC Basic waste buddle type 10: 1×2.3/3.2 mm Ext.Ø., 1×5 to 11.5 mm Int.Ø., 1×10 L canister, sieve and splash guard (76503-884)
HPLC Basic waste buddle type 20: 2×2.3/3.2 mm Ext.Ø., +5.0 to 11.5 mm Int.Ø., 1×20 L canister, with splash guard and change indicator (76503-994)

Certifications: UN Approved.

Delivery information: Supplied with: 1× Safety waste cap, 1× exhaust filter M.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR