Griffin Style, Low Form Borosilicate Glass Beakers with Spout
Supplier: Wards
BK0112 BK0107 BK0111 BK0110 BK0103 BK0104 BK0105 BK0102 BK0106 BK0108
470343-824EA 104.75 CAD
470343-824 470344-110 470344-270 470344-642 470344-660 470344-890 470344-906 470344-938 470344-956 470345-672
Griffin Style, Low Form Borosilicate Glass Beakers with Spout
Beakers
Pack of Griffin style glass beakers, perfect for precise measurements, lab experiments, and reliable mixing.
  • White graduations
  • Spout for easy pouring
  • Uniform walls
Eisco Labs Borosilicate glass beakers have the following heat tolerances: Strain Point −515 °C | Annealing Point 565 °C | Softening Point 820 °C.
Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.