Elastic Confined Space Covers
Supplier: Brady Worldwide
177438 177439 177440 177441 177442
77574-890EA 132.1 CAD
77574-890 77574-874 77574-892 77574-850 77574-918
Elastic Confined Space Covers
Signs
Get a compliant sign and barrier in one. Enhance visibility, reinforce compliance and keep unauthorized personnel out of confined spaces with an elastic confined space cover.
  • Washable, strong material offers long-lasting durability in various industrial and environmental conditions
  • Mesh construction helps prevent entry while allowing beneficial airflow
  • Bright colors and reflective elements enhance visibility
  • Compact and lightweight for easy transport and storage
  • Application: Lockout tagout
  • Attachment type: Elastic
  • Vented
Elastic construction enables cover to wrap cover around bolts, hinges and other cover accessories or obstructions with ease. DANGER sign is compliant with OSHA CFR 1910.146(c)(2) and cover helps prevent unauthorized entry for two-pronged safety.