Magnetic Confined Space Covers
Supplier: Brady Worldwide
177433 177434 177435 177436 177437
77574-888EA 179.61 CAD
77574-888 77574-866 77574-896 77574-940 77574-846
Magnetic Confined Space Covers
Signs
Get a compliant sign and barrier in one. Enhance visibility, reinforce compliance and keep unauthorized personnel out of confined spaces with a magnetic confined space cover.
  • Washable, strong material offers long-lasting durability in various industrial and environmental conditions
  • Mesh construction helps prevent entry while allowing beneficial airflow
  • Bright colors and reflective elements enhance visibility
  • Compact and lightweight for easy transport and storage
  • Application: Lockout tagout
  • Attachment type: Magnetic
  • Vented
Built-in magnets secure cover to metal surfaces without the need for fasteners or tools. DANGER sign is compliant with OSHA CFR 1910.146(c)(2) and cover helps prevent unauthorized entry for two-pronged safety.