L(+)-Histidine monohydrochloride monohydrate 98.5-101.0%, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

4938-06 4938-07 4938-09
MK4938-06EA 1874.28 CAD
MK4938-06 MK4938-07 MK4938-09
L(+)-Histidine monohydrochloride monohydrate 98.5-101.0%, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™
L(+)-Histidine monohydrochloride monohydrate
Formula: C₆H₉N₃O₂·HCl·H₂O
Melting Pt: 259 °C
Density: 1.485 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151027
CAS Number: 5934-29-2

Order Now

Specification Test Results

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets ChP Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets E.P. Chemical Specifications
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
EP/BP - Assay (C6H9N302 HCl) (dried basis) 98.5 - 101.0%
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Identification F Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 3.0 - 5.0
EP/BP - Specific Rotation (.A]2°AO (+) 9.2 - 10.6°
EP/BP-Ninhydrin-Positive Substances-Each ≤0.2%
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤0.5%
EP/BP - Sulfate (S04) ≤300 ppm
EP/BP - Ammonium (NH4) ≤0.02%
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Drying at 150 °C 7.0 - 10.0%
EP/BP - Ash (sulfated) ≤0.1%
JP - Identification Passes Test
JP - 0ptical Rotation (+) 9.2 - 10.6
JP - pH 3.5 - 4.5
JP - Clarity and Color of Solution Passes Test
JP - Sulfate (S04) ≤0.028%
JP - Ammonium (NH4) ≤0.02%
JP - Iron (Fe) ≤10 ppm
JP - Related Substances Passes Test
JP - Water 7.2 - 10.0%
JP - Residue on Ignition ≤0.10%
JP - Assay (anhydrous basis) 99.0 - 101.0%
ChP - Specific Rotation @ 20 °C 8.5 - 10.5°
ChP - Identification 1 (TLC) Passes Test
ChP - Identification 2 (IR) Passes Test
ChP - Acidity or alkalinity (pH) 3.5 - 4.5
ChP - Transmittance of solution ≥ 98.0%
ChP - Chloride Content 16.7 - 17.1%
ChP - Sulfates ≤0.02%
ChP - Ammonium ≤0.02%
ChP - 0ther amino acids (TLC)(<=0.2%, one spot only) Passes Test
ChP - Loss on drying @ 105 °C ≤0.2%
ChP - Residue on ignition ≤0.1%
ChP - Heavy metals ≤0.001%
ChP - Arsenic ≤0.0001%
ChP - Bacterial endotoxin, EU/g ≤6.0
ChP - Assay, dried basis 99.0 - 101.0%
ChP - Iron ≤0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR