L-Arginine hydrochloride 98.5-101.0% USP, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

4922-06 4922-07 4922-09
MK4922-06EA 1282.49 CAD
MK4922-06 MK4922-07 MK4922-09
L-Arginine hydrochloride 98.5-101.0% USP, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™
L-Arginine hydrochloride
Formula: C₆H₁₅ClN₄O₂
Melting Pt: 218 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064550
CAS Number: 1119-34-2

Order Now

Specification Test Results

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets ChP Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets E.P. Chemical Specifications
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C6H14N402·HCl) (dried basis) 98.5 - 101.5%
USP - Identification Passes Test
USP - Specific Rotation (A]2°AO (+) 21.4 - 23.6°
USP - Loss on Orying at 105 °C ≤0.2%
USP - Residue on Ignition ≤0.1%
USP - Sulfate (S04) ≤0.03%
USP - Related Compounds - Individual Impurities ≤0.5%
USP - Related Compounds - Total Impurities ≤2.0%
USP - Chloride (Cl) 16.5 - 17.1%
EP/BP - Assay (C6H14N402·HCl) (dried basis) 98.5 - 101.0%
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification E Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation (A]2°AO (+) 21.0 - 23.5°
EP/BP-Ninhydrin-Positive Substances-Each ≤0.2%
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤0.5%
EP/BP - Sulfate (S04) ≤300 ppm
EP/BP - Ammonium (NH4) ≤0.02%
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Orying ≤0.5%
EP/BP - Ash (sulfated) ≤0.1%
JP - Assay (C6H14N402·HCl) (dried basis) 98.5 - 101.0%
JP - Identification Passes Test
JP - 0ptical Rotation (+) 21.5 - 23.5°
JP - pH (1 in 10) 4.7 - 6.2
JP - Clarity and Color of Solution Passes Test
JP - Sulfate (S04) ≤0.028%
JP - Ammonium (NH4) ≤0.02%
JP - Arsenic (As) ≤2 ppm
JP - Related Substances Passes Test
JP - Loss on Orying at 105 °C ≤0.20%
JP - Residue after Ignition ≤0.10%
Endotoxin Concentration, IU/mg, For Information only
ChP - Transmittance of solution ≥ 98.0%
ChP - Sulfates ≤0.02%
ChP - Ammonium ≤0.02%
ChP - Loss on drying @ 105 °C ≤0.2%
ChP - Residue on ignition ≤0.1%
ChP - Heavy metals ≤0.001%
ChP - Arsenic ≤0.0001%
ChP - Identification 1 (TLC) Passes Test
ChP - Identification 2 (IR) Passes Test
ChP - Chloride Content 16.5 - 17.1%
ChP - 0ther amino acids (TLC)(<=0.2%, one spot only) Passes Test
ChP - Proteins Passes Test
ChP - Iron ≤0.001%
ChP - Specific Rotation @ 20 °C (+) 21.5 - 23.5
ChP - Phosphates ≤0.02%
ChP - Assay (dried basis) 98.5 - 101.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR