Cohesive Slot Samplers
Supplier: SAMPLING SYSTEMS LTD
2210A-1000 2210A-600
77576-364EA 2567.88 CAD
77576-364 77576-368
Cohesive Slot Samplers
Samplers Solid Samplers
The cohesive slot sampler is designed for taking cross sectional samples of cohesive powders.
  • Made from 316L stainless steel
  • Can be completely dismantled for quick and thorough cleaning
  • Available from stock
  • Fast delivery
The cohesive slot sampler is excellent for taking cross sectional samples from powders with poor flow characteristics. Upon rotating the device, the scraper pushes powder into the sampler. When finished, the sampler can be compeltely dismantled for quick and easy cleaning.