Cohesive Pocket Samplers
Supplier: SAMPLING SYSTEMS LTD
2220B-600 2220B-900
77576-372EA 2019.84 CAD
77576-372 77576-374
Cohesive Pocket Samplers
Samplers Solid Samplers
The cohesive pocket sampler is designed for taking discreet samples of cohesive powders.
  • Ingenious scraper pushes powder into containment when rotated
  • Made from 316L stainless steel
  • Available from stock
  • Fast delivery
The cohesive pocket sampler is perfect for taking discreet samples of powders with poor flow characteristics. The scraper pushes powder into the sampler when rotated, while a solid inner rod ensures that samples are kept separate from each other.