PowderThief Single Use Samplers
Supplier: SAMPLING SYSTEMS LTD
8060H-1001S 8060H-1005S 8060H-1010S 8060H-501S 8060H-505S 8060H-520
77576-910EA 469.12 CAD
77576-910 77576-914 77576-918 77576-930 77576-938 77576-944
PowderThief Single Use Samplers
Samplers Disposable Samplers
Disposable samplers specially designed for powders and granules.
  • No risk of cross-contamination
  • Assembled in a cleanroom environment
  • Economical - save both time and money
Suitable for industries such as pharmaceutical, food, chemical, cosmetic, biotechnology. Pre-sterilised versions are available.
Certifications: FDA conformity. BSE/TSE free. Full batch traceability, ATEX rated.
Packaging: Each sampler is wrapped in a plastic bag.