UVP ChemSolo Auto Imager
Supplier: Analytik Jena CA
849-97-0948-01 849-97-0948-02 849-97-0948-03 849-97-0948-04
CA77574-788EA 39535 CAD
CA77574-788 CA77574-790 CA77574-792 CA77574-794
UVP ChemSolo Auto Imager
Photodocumentation Systems
The UVP ChemSolo auto imager - a versatile bioimaging system designed to streamline a wide range of applications that can go from capturing fluorescent DNA and protein gels to analyzing chemiluminescent western blots and facilitating colony counting.
  • High resolution chemiMOS camera at 9 MP capability
  • Three-position automated filter tray for easy switching between emission filters
  • Broad-band emission filter is included with the unit
  • Overhead epi-white and epi-blue LED lights enable a wide range of applications
  • Wide front door with UV protection; a safety switch automatically turns off the UV light when the door is open
  • UVP Elite UV transilluminator offers an illumination size of 16.8×21 cm with a maximum field of view of 16×16 cm
A versatile bioimaging system designed to streamline a wide range of applications that can go from capturing fluorescent DNA and protein gels to analyzing chemiluminescent western blots and facilitating colony counting. Its intuitive automation and seamless integration with VisionWorks® Software empower users with comprehensive control over image acquisition and analysis, all conveniently accessible from their computer screens.
VisionWorks® Software offers intuitive capture controls and advanced image enhancement and analysis tools, streamlining the process for users. It provides a comprehensive solution for efficiently managing sample images, from auto and manual image capture modes to customizable exposure settings and image saving options (TIFF or JPEG).
Certifications: CSA/CE compliant.