VWR® Disposable Cleanroom Sterile Sleeves, Made in USA
Supplier: Avantor
VWR®
653-S
76944-184CS 722 CAD
76944-184
VWR® Disposable Cleanroom Sterile Sleeves, Made in USA
Disposable Garments
Premium ultrasonically welded disposable sterile sleeves are crafted for cleanroom environments. Made of 3-layer SMS polypropylene with a 44gsm weight, they provide a secure fit with elastic wrist and top cuffs.
  • Ultrasonic welded encapsulated seam construction
  • 3-layer SMS polypropylene
  • 44 gsm weight
  • 24" length, 9.5" width
  • Specialized arm entry for easy donning
  • Gamma sterilized for single-use cleanroom applications
  • Manufactured according to ISO 13485 guidelines
Premium ultrasonically welded disposable sterile sleeves are crafted for cleanroom use, and feature a 3-layer material composition made of SMS polypropylene. The elastic components include a ¹/₄" wrist cuff and a ¹/₈" top cuff for a secure fit. The top cuff is folded back 9", with an open-end arm entry design to facilitate easy donning.
These sleeves are gamma sterilized for single-use applications. Each unit is packaged individually. Made in the USA, with all components manufactured domestically.
Packaging: Sleeves are packaged individually in an Oxygen barrier peel bag, (poly-lined carton). 200 sleeves per case.