Formamidine hydrobromide, low water content ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

F0973-1G F0973-5G
TCF0973-1GEA 39.5 CAD
TCF0973-1G TCF0973-5G
Formamidine hydrobromide, low water content ≥98.0% (by total nitrogen and titration analysis)
Formamidine hydrobromide
Formula: CH₄N₂·HBr
MDL Number: MFCD28369274
CAS Number: 146958-06-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Water max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR