Monarch® Mag Viral DNA/RNA Extraction Kits

Supplier: New England Biolabs (NEB)

T4010L T4010S T4010X
CA77532-266EA 1313 CAD
CA77532-266 CA77532-264 CA77532-268
Monarch® Mag Viral DNA/RNA Extraction Kits
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

The Monarch Mag Viral DNA/RNA Extraction Kit provides a rapid and reliable magnetic bead-based process for extracting viral nucleic acids from saliva and respiratory swab samples. The kit combines the efficiency of silica-based nucleic acid purification with the ease of use of magnetic beads.


  • Designed for hands-free extraction of viral DNA and/or RNA
  • Tested for saliva and respiratory swab sample types. Compatible with wastewater samples, after enrichment steps (not supplied)
  • Suitable for qPCR/RT-qPCR, ddPCR, library prep for sequencing/NGS and other downstream applications
  • Includes carrier RNA for sensitive detection in RNA-based amplification workflows


Utilizes magnetic bead-based methods. Compatible with manual and automated high-throughput workflows on a variety of instrument platforms, such as KingFisher® Flex, Agilent® Bravo®, MGISP® liquid handlers, and more.


Processing format: Manual or automated
Sample purification: Viral DNA and RNA* from respiratory viruses (enveloped and non-enveloped, dsDNA and ssRNA)
Carrier supplier: Poly A carrier RNA
Elution volume: 33 to 100 μl
Tested automation platforms: KingFisher Flex; Agilent Bravo and MGISP liquid handlers


Caution: For Research Use Only. Not for use in diagnostic procedures.

New Product

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Target molecule
Refine target molecule
Sample type Saliva,Throat swab specimens,Nasal and nasopharyngeal swab specimens
Method / Format Magnetic beads
Sample size Up to 200 μl
Binding capacity Upto 3 μg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR