Monarch® Spin Collection Tubes
Supplier: New England Biolabs (NEB)
Monarch® Spin collection tubes are designed to be used with Monarch spin columns.
- Compatible with all Monarch® spin columns
- Designed and manufactured with less plastic than other leading suppliers
Monarch® Spin Collection Tubes are designed to be used with Monarch® Spin Columns for collection of waste and other fractions during centrifugation steps in Monarch® Spin Nucleic Acid Purification Kits.
Ordering information: Available for purchase separately. Included with Monarch Spin nucleic acid purification kits.
Packaging: Supplied as 100 collection tubes, in 2 packs of 50 tubes each.