Recombinase Polymerase Amplification (RPA) Kit
Supplier: G-Biosciences
786-2155
786-2156
786-2157
77540-368EA
499.99
CAD
77540-368
77540-370
77540-372
Recombinase Polymerase Amplification (RPA) Kit
Nucleic Acid Reagents
Isothermal Amplification Reagents
A mixture of RPA enzymes (UvsX, UvsY, GP32 and Bsu) in liquid form.
- Use for isothermal amplification
Rapid diagnostic assay.