Anti-TER119 Rat Monoclonal Antibody [clone: TER-119] (FITC)

Supplier: STEMCELL Technologies
100-0453 100-0454
MSPP-100-0453EA 161.54 CAD
MSPP-100-0453 MSPP-100-0454
Anti-TER119 Rat Monoclonal Antibody [clone: TER-119] (FITC)
Antibodies
Rat (WI) monoclonal IgG2b antibody against mouse TER119.

  • Erythroid cells
  • Red blood cells

The TER-119 antibody reacts with murine TER119 (Ly-76), an ~52 kDa protein associated with glycophorin A on the surface of cells of the erythroid lineage in embryonic yolk sac, fetal and newborn liver, adult bone marrow, peripheral blood, and lymphoid organs. TER119 is an erythroid-specific marker expressed at all stages of differentiation from early proerythroblasts to mature erythrocytes, but not by erythroid colony-forming cells (BFU-E, blast-forming unit erythroid, or CFU-E, colony-forming unit erythroid). The TER-119 antibody is a component of the "lineage cocktail" used to detect, or deplete cells committed to hematopoietic lineages. In adult mice, TER119 is found on ~20 - 25% of bone marrow cells and ~2 - 3% of splenocytes. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD4+ T Cell Isolation Kit (Catalog #19852).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: TER119
Clonality: Monoclonal
Clone: TER-119
Conjugation: FITC
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR