Anti-Beta-tubulin III Mouse Monoclonal Antibody [clone: AA10]

Supplier: STEMCELL Technologies
60100 60100.1
MSPP-60100EA 775.38 CAD
MSPP-60100 MSPP-601001
Anti-Beta-tubulin III Mouse Monoclonal Antibody [clone: AA10]
Antibodies
Mouse monoclonal IgG2a antibody against human, mouse, rat beta-tubulin III, unconjugated.

  • Neural cells Psc derived
  • Neurons

The AA10 antibody reacts with beta-tubulin III, an ~50 - 55 kDa structural protein that is a constituent of tubulin. Tubulin is the major component of microtubules within the cytoskeleton and is assembled from heterodimers of alpha and beta tubulin subunits. The beta III isoform of tubulin, also known as neuron-specific class III beta-tubulin, is expressed primarily in neurons and is widely used as a marker to distinguish neurons from other cell types. Beta-tubulin III contributes to microtubule formation in neuronal cell bodies and axons, a function involving GTP binding, and plays roles in axonal transport, neuronal cell proliferation, and differentiation. It is highly expressed in several types of cancer and is a predictive and prognostic marker for various tumors, for example, being found in neoplastic but not in normal glial cells. The AA10 antibody is expected to recognize all mammalian homologs of beta-tubulin III.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: Beta-tubulin III
Clonality: Monoclonal
Clone: AA10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR