Gram's iodine solution Avanced
Supplier: G-Biosciences
Synonyms:
Lugol's iodine, Iodine and potassium iodide, Potassium triiodide
786-2097
786-2098
77540-252KT
175.76
CAD
77540-252
77540-254
Gram's iodine solution Avanced
Gram's iodine solution
Gram Iodine Advanced Solution acts as a mordant .
- An aqueous solution of Iodine
It is used in Gram staining to differentiate gram-positive from gram-negative bacteria
Formula:
I₃K MW: 419.812 g/mol |
MDL Number:
MFCD00134128 CAS Number: 12298-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...