Gram's iodine solution PVP

Supplier: G-Biosciences

786-2095 786-2096
77540-248EA 564.39 CAD
77540-248 77540-250
Gram's iodine solution PVP
Gram's iodine solution

Gram Iodine PVP Solution acts as a mordant .


  • An aqueous solution of PVP Iodine


It is used in Gram staining to differentiate gram-positive from gram-negative bacteria

Formula: I₃K
MDL Number: MFCD00134128
CAS Number: 12298-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR