Gram's decolourizing solution Advanced
Supplier: G-Biosciences
Synonyms:
Gram's Decolorizer Solution, Gram's Decoloriser Solution
786-2088
786-2089
77540-234EA
316
CAD
77540-234
77540-236
Gram's decolourizing solution Advanced
Gram's decolorizing solution
Gram Decolorizer is a mixture of 25% Acetone and 75% Isopropanol solution.
- An aqueous solution of Acetone and Isopropanol
It is used for rapid decolorization before counterstaining