Gram′s safranin solution Advanced

Supplier: G-Biosciences

Synonyms: Safranin T solution

786-2103 786-2102
77540-264KT 226.21 CAD
77540-264 77540-262
Gram′s safranin solution Advanced
Gram′s safranin solution

Gram Stain Safranin, Advanced acts as a counterstain .


  • An aqueous dye solution


It is used in Gram staining to differentiate gram-positive from gram-negative bacteria

Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
MDL Number: MFCD00011759
CAS Number: 477-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR