Human/Mouse Recombinant GDF-11
Supplier: STEMCELL Technologies
78110 78110.1
MSPP-78110EA 429.23 CAD
MSPP-78110 MSPP-781101006
Human/Mouse Recombinant GDF-11
Proteins and Peptides
Growth differentiation factor 11 (GDF-11) is a member of transforming growth factor beta (TGF-β) family. It binds to the TGF-β receptors ALK4, ALK5, and ALK7 and activates the SMAD signaling pathway (Ho et al.). GDF-11 regulates the development of the olfactory system, retina, and pancreas, as well as anterior/posterior patterning of the axial skeleton (Lee and Lee). GDF-11 is an endocrine factor expressed in skeletal muscle, brain, and dental pulp (Kondás et al.). Studies in mice showed that GDF-11 regulates muscle and cardiac aging, and stimulates neurogenesis by remodeling blood vessels (Katsimpardi et al.; Loffredo et al.; Sinha et al.). This product is animal component-free.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').