EnGen SpRY Cas9
Supplier: New England Biolabs (NEB)
M0669M M0669T
CA77532-248EA 1004 CAD
CA77532-248 CA77532-250
EnGen SpRY Cas9
Enzymes
EnGen® SpRY Cas9 from Streptococcus pyogenes is an engineered, RNA-guided, DNA endonuclease that catalyzes site-specific cleavage of double-stranded DNA (dsDNA).
  • Unconstrained DNA-targeting capabilities (5'-NNN-3' PAM)
  • Ideal for digestion of large plasmids in cloning workflows
Eliminate sequence constraints for dsDNA targeting with non-specific PAM (5´-NNN-3´ PAM). Use in conjunction with the EnGen sgRNA synthesis kit, S. pyogenes, EnGen mutation detection kit, and NEBuilder HiFi DNA assembly master mix.