LyoPrime WarmStart™ Fluorescent LAMP/RT-LAMP Mixes (with UDG)
Supplier: New England Biolabs (NEB)
L4401P L4401S L4401S
CA77532-240EA 584 CAD
CA77532-240 CA77532-242 77532-242
LyoPrime WarmStart™ Fluorescent LAMP/RT-LAMP Mixes (with UDG)
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Fluorescent LAMP/RT-LAMP mix is designed to provide a simple one-step solution for Loop-Mediated Isothermal Amplification (LAMP) of DNA or RNA (RT-LAMP) targets.
  • Reduce risk of carryover contamination with thermolabile UDG and dUTP included in the master mix
  • WarmStart technology inhibits activity at room temperature
  • Simple, rapid rehydration
Lyophilized for stable shipping and storage at ambient/room temperature. Can be stored at room temperature for up to two years before rehydration. Contains fluorescent dye for real-time fluorescence measurement during LAMP/RT-LAMP reactions.