Micro-Touch® DermaClean® Examination Gloves, Latex
Supplier: Ansell Healthcare
107425 107426 107427 107428 107429
77531-232BX 16.9 CAD
77531-232 77531-236 77531-230 77531-234 77531-238
Micro-Touch® DermaClean® Examination Gloves, Latex
Gloves
These ambidextrous gloves are fully textured and therefore provide an excellent grip in wet and dry conditions. They are ideal for instrumentation, handling test tubes or infusion equipment in labs, pharmaceutical environments and general wards.
  • Latex, powder-free medical
  • For an excellent grip in wet and dry conditions
  • For easy donning
  • Provides great comfort and flexibility
  • Secure grip
  • Comfortable fit and easy donning
  • Freedom from holes: 1.5 AQL
Recommended for brief, low-risk examination procedures.
Certifications: Certified of EN ISO 21420:2020, ISO 10993-1, ISO 11193-1:2008, ISO 10993-5 and ISO 10993-10:2002. ISO 9001, ISO 13485 and ISO 10993-10.
Caution: This product contains natural rubber latex, which may cause allergic reactions.