Cleanroom Sticky Notes
Supplier: Columbia Printing & Graphics
POST-IT-CAN POST-IT-GRE
77574-766CS 219.46 CAD
77574-766 77574-778
Cleanroom Sticky Notes
Notepads
Class 10 cleanroom sticky notes include 100 latex sheets per pad and are designed for use in critical environments where low particulation and ESD's are required.
  • 28# latex paper
  • 3×3" note pads
  • 24 pads per case
The latex sheets are smudge-resistant and the adhesive used for binding the sheets does not emit ESD's or carry particulates when a sheet is removed.