Floor Mounted Service Sinks, Advance Tabco®
Supplier: Advance Tabco
9-OP-48 9-OP-20
47747-440EA 6922.38 CAD
47747-440 58003-086
Floor Mounted Service Sinks, Advance Tabco®
Faucets and Sinks Sinks
Floor mounted unit eliminates the need for lifting heavy containers.
The sink bowl is deep drawn for increased capacity and features one-piece construction of heavy gauge, Type 304 stainless steel. Available in two bowl dimensions.
Ordering information: Faucet not included; contact your Avantor Furniture Specialist for more information.
Order Now
SPECIFICATIONS
Environmentally Preferable