TX®728 Pre-wetted Swabs
Supplier: Texwipe
TX®728
TX728
77547-534CS 1570.63 CAD
77547-534
TX®728 Pre-wetted Swabs
Swabs Swabs for Cleaning
TX®728 Pre-wetted swabs with 100% Methanol solution from 100% polyester hydroentangled nonwoven material. Complete thermal bond construction eliminates adhesive contamination or head material releasing from the swab handle.
  • Double layer of polyester hydroentangles nonwoven fabric for softness and solvent capacity
  • 100% virgin polypropylene handle ensures no additional contaminants are introduced while offering excellent chemical resistance
  • Pre-wet with 0.2 μm filtered 100% HPLC-grade methanol for ease of use and prevents need of solvent containers and difficulty of wetting swab head
  • Replaces practice of wetting a wiper or swab with cleaning solution which may provide inconsistent cleaning results
  • Pre-wetted swabs provide a consistent wetness level for optimized cleaning efficiency and repeatable VOC levels. No free liquid in pouch
  • Swab is designed for use in hard to reach places
  • Fast evaporation time with no residual residue
Cleanroom manufactured, made to exacting and consistent tolerances using high-precision automated processes. Lot coded for traceability and quality control. Swabs are packaged in pairs to ensure a lot of clean capacity, reducing solvent evaporation that preserves a consistent swab wetness level and solvent exposure.
Trademarked light-green colored handles with 'TEXWIPE' name embossed on handle.
Precision spot cleaning of optical components such as lenses, mirrors, or other surfaces.
Used in aerospace, data storage, laboratory, industries, microelectronics, optics and semiconductors.
Certifications: ISO 9001:2015.
Packaging: Packaged in a metal foil single use pouches silicone-free and amide-free bag. Outer packaging of red slider bag to easily identify and store pouches. GHS labeling on back of red slider bag.