Space Saver Hand Sink, Advance Tabco®
Supplier: Advance Tabco
7-PS-23
58003-098EA 3573.03 CAD
58003-098
Space Saver Hand Sink, Advance Tabco®
Furniture Casework
Space saving design is only 30.5 cm (12") wide.
  • Constructed of heavy gauge, Type 304 stainless steel
Unit has a deep drawn bowl for greater capacity. It has a splash mounted 10.2 cm (4") spread gooseneck faucet.