Square Corner Scullery Budget Sinks, Advance Tabco®
Supplier: Advance Tabco
4-3-36 4-1-24
58003-076EA 5287.93 CAD
58003-076 58003-078
Square Corner Scullery Budget Sinks, Advance Tabco®
Furniture Casework
All heli-arc welded sinks are constructed of 16-gauge, type 304 stainless steel.
  • Available in one or three compartment styles
Galvanized tubular legs have adjustable feet.
Ordering information: Faucet should be ordered seperately.
Delivery information: Unit supplied with 3.8 cm (1¹/₂") IPS basket drain.
Order Now
SPECIFICATIONS
Environmentally Preferable