PYREX® Kuderna-Danish Concentrator Apparatus, [ST] Joints, Corning
Supplier: Corning
2158-500 2158-250
89091-412CS 864.62 CAD
89091-412 89091-616
PYREX® Kuderna-Danish Concentrator Apparatus, [ST] Joints, Corning
Concentrators
The complete PYREX® Kuderna-Danish concentrator apparatus consists of a 3-ball Snyder column with 24/40 joints, a flask with a 24/40 outer upper joint and 19/22 inner lower joint, and a graduated tube with 19/22 outer joint. If further concentration is desired, select a 2-ball column (2158-2CO) and combine with the Kuderna-Danish tube of your choice.
  • The 250 and 500 ml complete apparatus include a 10 ml tube
  • The 1000 ml complete apparatus includes a 25 ml tube
  • Apparatus is held together using glass hooks and 1" stainless steel springs
  • Standard taper botton
More Product Information