Epoxy Resin Tub Sinks, Kewaunee®
Supplier: Kewaunee
VF-1003-00 VF-1003-DT VF-1007-00 1008-00 1022-00 1027-00
VF-1003-00EA 1108.13 CAD
VF-1003-00 VF-1003-DT VF-1005-00 VF-1006-00 VF-1007-00 VF-1008-00 VF-1021-00 VF-1022-00 VF-1027-00
Epoxy Resin Tub Sinks, Kewaunee®
Furniture Casework
Tub sinks are made from a combination of modified epoxy resins and inert fillers to produce a durable, long-lasting material.
  • Formed in metal molds, the seamless, one-piece sinks feature coved corners and bottoms pitched to the 3.8 cm (1¹/₂") sized drain outlets
Ordering information: Available with one or two basins.