Disposable Coveralls, Extra Protection
Supplier: O&M HALYARD CANADA, INC CA
10075 10085 10095 10096
77531-648CS 297.36 CAD
77531-648 77531-650 77531-652 77531-654
Disposable Coveralls, Extra Protection
Disposable Garments
Disposable coverall extra protection.
  • Back gather
  • Offers protection from fluid strikethrough for light to moderate fluid contact
  • Three layer SMS fabric