Magicard® YMCKOK Color Ribbon
Supplier: Essendant
Magicard®
SRXMA250YMCKOK
77531-630EA 349.58 CAD
77531-630
Magicard® YMCKOK Color Ribbon
Printer Ribbons
Magicard® YMCKOK ribbons are ideal for dual-sided card printing with YMC, the first K panel, and O panel printing on the front side of the card and the second K panel printing on the back side of the card.
  • Full color six-panel YMCKOK dye film for dual-sided printing
  • Print full-color images on the front and one-color text or barcodes on the back
  • Features drop-in dye film loading which provides hassle-free installation
  • Estimated print yield of 250 individual single-sided card prints
Magicard ® consumables are specially designed, tested and approved for Magicard® printers to ensure high quality images and text. Genuine Magicard® consumables offer the best performance and extended lifespan of your card printer.