BRAND Transferpettor® Single-Channel Positive Displacement Pipettors, Variable Volume, BrandTech
Supplier: Brandtech
702804 702806 702810 702812
18900-086EA 1264.11 CAD
18900-086 18900-088 18900-090 18900-092
BRAND Transferpettor® Single-Channel Positive Displacement Pipettors, Variable Volume, BrandTech
Pipettes Micropipettes
These pipettors are the solution for many liquids that cannot be pipetted accurately with an air displacement pipet, such as viscous, volatile, or foaming liquids.
  • Instruments, tips, and seals are not autoclavable
  • Optional Transferpettor station holds two instruments
  • Tips and seals for convenient access in the workplace
Pipettors are useful when working with liquids such as concentrated protein solutions, oils, fats, resins, and glycerin. They can be used to accurately pipette surfactants and are designed for use with high-vapor pressure media, such as alcohol, acetone, and hydrocarbons.
The optional Transferpettor station holds two instruments, tips, and seals for convenient access in the workplace.
The tightly fitting piston comes in direct contact with liquid for reproducible results, without calibration adjustments. Tip walls are wiped clean to the last drop, for repeated use of the tips without cross-contamination.
Ordering information: Supplied with manufacturer’s one-year warranty. Pipettors are supplied with one tip and seal, seal mounting/removal tool, performance certificate, and operating manual.