VWR® Single-Channel Digital Microdispensers, Variable Volume
Supplier: VWR International
53506-100 53506-121 53506-143 53506-201 53506-223
53506-100EA 1753.37 CAD
53506-100 53506-121 53506-143 53506-201 53506-223
VWR® Single-Channel Digital Microdispensers, Variable Volume
Pipettes Micropipettes
This adjustable dispenser features a digital readout, and a positive displacement piston ensures complete recovery of the sample.
  • Accurate dual-calibration system
  • Easy to set and easy to use
  • Positive displacement pipetting of any fluid
  • Adjustable
  • Precision-molded plastic or glass capillary bores
A unique dual-calibration system provides an accuracy and reproducibility of better than ±1%.
Capillary bores are made of glass except for the 100 to 1000 µl volume and 1000 µl volume models, which are made of precision-molded plastic. Users can set wholeMore Product Information