Finnpipette® Novus Single-Channel Electronic Pipettors, Variable Volume, Thermo Scientific
Supplier: Thermo Fisher Scientific
46200000 46200200 46200600 46200800 46200300 46200400 46200500 46200700
89082-224EA 2233.43 CAD
89082-224 89082-226 89082-228 89082-236 89082-240 89082-230 89082-232 89082-234 89082-238
Finnpipette® Novus Single-Channel Electronic Pipettors, Variable Volume, Thermo Scientific
Pipettes Micropipettes
Pipettors combine simplicity, comfort, and functionality in an exceptionally intuitive instrument to improve productivity, precision, and ergonomic operation in liquid handling applications.
  • Lightweight design keeps stress and exertion to a minimum
  • Feature ten available pipetting functions and nine pipetting speeds
  • Designed for easy in-lab calibration and servicing, and come
  • Equipped with a fully autoclavable tip cone to ensure sterility and avoid cross-contamination
The user interface has a two-level intuitive menu for simple control, and is extremely easy-to-use thanks to its fully graphical text display. A multilingual graphical interface is offered in seven languages: English, French, German, Italian, Japanese, Spanish, and Swedish. Unique backlight technology distinguishes the display by eliminating surrounding light reflections and improving contrast. The result is a graphical display with excellent clarity, even in low light conditions such as safety cabinets and fume hoods. A lightweight, long life battery can be fully recharged in only one hour, while a long, unobtrusive cable permits use of the instrument during recharging. Pipettors can even be personalized with their own name, which is displayed on the screen when the instrument is not in use.
Advanced lightweight construction helps to prevent upper body stress and strain, allowing users to work over extended periods without fatigue. Ergonomic features include a solid handle shaped to fit comfortably and securely in the hand and a finger rest that can adjust over 120° for optimal positioning.