Acura® Manual Pipettor Triopacks, DWK Life Sciences
Supplier: DWK Life Sciences
Acura®
W810323 W810327 W810320 W810324 W810322 W810326 W810321 W810325 W810319
89044-610CS 2091.62 CAD
89044-610 89044-608 89044-604 75800-380 89044-606 89044-600 89044-612 89044-602 89044-614
Acura® Manual Pipettor Triopacks, DWK Life Sciences
Pipettes Micropipettes
Each ready-to-use set includes three manual adjustable pipettors and accessories.
Nine different specific volume combinations are available with full coverage ranging from 0.1 µl to 10 ml.
Ordering information: Each set also includes tip samples, QC certificate, and operating instructions.